5-Fluoro-2-nitrotoluene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Fluoro-3-methyl-4-nitrobenzene

F0301-5G F0301-25G
TCF0301-5GEA 28 USD
TCF0301-5G TCF0301-025G
5-Fluoro-2-nitrotoluene ≥98.0%
5-Fluoro-2-nitrotoluene

CAS Number: 446-33-3
MDL Number: MFCD00007283
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 98
Flash Point (°C): 88
Freezing point (°C): 23

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 98 °C
Melting Pt: 28 °C
Density: 1.27 g/cm³ (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00007283
CAS Number: 446-33-3
EINECS: 207-164-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b932157ec53a433adcf10117bb40a1aac5d767c9&vid=f2d2edeb060b9a548cd3ada10389eef7d12ec9b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR