(3-Fluorophenyl)acetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 2-(3-Fluorophenyl)acetonitrile, 3-Fluorobenzyl cyanide

F0256-5G F0256-25G
TCF0256-5GEA 35 USD
TCF0256-5G TCF0256-025G
(3-Fluorophenyl)acetonitrile ≥98.0%
(3-Fluorophenyl)acetonitrile

CAS Number: 501-00-8
MDL Number: MFCD00001907
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 230
Flash Point (°C): 113
Freezing point (°C): 21


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluorophenylacetonitrile

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 230 °C
Density: 1.14058 g/cm³ (15 °C)
Flash Pt: 113 °C
MDL Number: MFCD00001907
CAS Number: 501-00-8
EINECS: 207-918-2
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64089203d67fb3dd579d37d6a261b2b6d427e759&vid=64089203d67fb3dd579d37d6a261b2b6d427e759&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR