2-Furoyl chloride ≥98.0%

Supplier: TCI America

F0233-500G F0233-25G
TCF0233-500GEA 337 USD
TCF0233-500G TCF0233-25G
2-Furoyl chloride ≥98.0%
2-Furoyl chloride

CAS Number: 527-69-5
MDL Number: MFCD00003228
Molecular Formula: C5H3ClO2
Molecular Weight: 130.53
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Melting point (°C): -2
Flash Point (°C): 85
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: 2-Furancarbonyl Chloride

Formula: C₅H₃ClO₂
MW: 130.53 g/mol
Boiling Pt: 174 °C
Melting Pt: –2 °C
Density: 1.33 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00003228
CAS Number: 527-69-5
EINECS: 208-422-9
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc011f14705560fa3be3d3e04554565dfde0163&vid=5fc011f14705560fa3be3d3e04554565dfde0163&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR