9-Hydroxyfluorene ≥99.0%
Supplier: TCI America
Synonyms:
9-Fluorenol, Fluoren-9-ol
F0229-250G
F0229-25G
TCF0229-250GEA
524.44
USD
TCF0229-250G
TCF0229-025G
9-Hydroxyfluorene ≥99.0%
9-Hydroxyfluorene
CAS Number: 1689-64-1
MDL Number: MFCD00001135
Molecular Formula: C13H10O
Molecular Weight: 182.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 155
Other Product Names / Synonyms: 9-Hydroxyfluorene
Formula:
C₁₃H₁₀O MW: 182.22 g/mol Boiling Pt: 320 °C (1013 hPa) Melting Pt: 155 °C Density: 1.151 g/cm³ (20 °C) |
MDL Number:
MFCD00001135 CAS Number: 1689-64-1 EINECS: 216-879-0 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...