4-Fluorobenzonitrile ≥99.0%

Supplier: TCI America
Warning

Synonyms: p-Fluorobenzonitrile

F0221-500G F0221-25G F0221-100G
TCF0221-500GEA 244 USD
TCF0221-500G TCF0221-025G TCF0221-100G
4-Fluorobenzonitrile ≥99.0%
4-Fluorobenzonitrile

CAS Number: 1194-02-1
MDL Number: MFCD00001812
Molecular Formula: C7H4FN
Molecular Weight: 121.11
Purity/Analysis Method: >99.0% (GC)
Form: Lump
Boiling point (°C): 188
Flash Point (°C): 65
Freezing point (°C): 34

Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 188 °C
Melting Pt: 37…40 °C
Density: ~ 1 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00001812
CAS Number: 1194-02-1
EINECS: 214-784-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4883016d1205d99d67b6717915808382498b0c03&vid=14c45d04bd60229a615afe639d981e2bfc4824d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR