4-Fluorobenzoyl chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: p-Fluorobenzoyl chloride

F0213-500G F0213-25G F0213-100G
TCF0213-500GEA 216 USD
TCF0213-500G TCF0213-25G TCF0213-100G
4-Fluorobenzoyl chloride ≥97.0% (by titrimetric analysis)
4-Fluorobenzoyl chloride

CAS Number: 403-43-0
MDL Number: MFCD00000684
Molecular Formula: C7H4ClFO
Molecular Weight: 158.56
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 82
Freezing point (°C): 10
Specific Gravity (20/20): 1.32

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 193 °C
Melting Pt: 9 °C
Density: 1.32 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000684
CAS Number: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac778407ad243011fce6218904b88ea3773521a7&vid=ac778407ad243011fce6218904b88ea3773521a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR