1-Fluoronaphthalene ≥98.0%
Supplier: TCI America
|
Warning
|
F0212-25G
F0212-5G
TCF0212-025GEA
45.79
USD
TCF0212-025G
TCF0212-005G
1-Fluoronaphthalene ≥98.0%
1-Fluoronaphthalene
CAS Number: 321-38-0
MDL Number: MFCD00003873
Molecular Formula: C10H7F
Molecular Weight: 146.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 65
Specific Gravity (20/20): 1.14
Formula:
C₁₀H₇F MW: 146.16 g/mol Boiling Pt: 212 °C Melting Pt: –12.5 °C Density: 1.14 (20 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003873 CAS Number: 321-38-0 EINECS: 206-287-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...