4-Fluorobenzyl chloride ≥98.0%

Supplier: TCI America
Danger

Synonyms: 4-Fluorobenzyl chloride, p-Fluorobenzyl chloride, 1-Chloromethyl-4-fluorobenzene, α-Chloro-4-fluorotoluene, 1-(Chloromethyl)-4-fluorobenzene

F0204-500G F0204-25G
TCF0204-500GEA 375 USD
TCF0204-500G TCF0204-25G
4-Fluorobenzyl chloride ≥98.0%
4-Fluorobenzyl chloride

CAS Number: 352-11-4 MDL Number: MFCD00000913 Molecular Formula: C7H6ClF Molecular Weight: 144.57 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 185 Flash Point (°C): 60 Specific Gravity (20/20): 1.21


Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 60
Specific Gravity (20/20): 1.21

Formula: C₇H₆ClF
Boiling Pt: 185 °C
Melting Pt: -18 °C
Density: 1.21 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000913
CAS Number: 352-11-4
EINECS: 206-516-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f24c0f2e9018d863f94560d36e7d01bcdcd65e03&vid=61829482a0e81b43ca197777c3965bf7a81be84a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR