2-Fluorobenzoyl chloride ≥98.0%

Supplier: TCI America
Danger

Synonyms: o-Fluorobenzoyl chloride

F0195-500G F0195-25G
TCF0195-500GEA 153 USD
TCF0195-500G TCF0195-25G
2-Fluorobenzoyl chloride ≥98.0%
2-Fluorobenzoyl chloride

CAS Number: 393-52-2
MDL Number: MFCD00000656
Molecular Formula: C7H4ClFO
Molecular Weight: 158.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 82
Specific Gravity (20/20): 1.34

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 206
Density: 1.34 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000656
CAS Number: 393-52-2
EINECS: 206-887-2
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e31175a4a574e00c34f80bcc90aeebadcc7cd544&vid=e31175a4a574e00c34f80bcc90aeebadcc7cd544&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR