4-Fluorobenzaldehyde ≥98.0%

Supplier: TCI America

F0191-25ML F0191-250ML
TCF0191-025MLEA 41.5 USD
TCF0191-025ML TCF0191-250ML
4-Fluorobenzaldehyde ≥98.0%
4-Fluorobenzaldehyde

CAS Number: 459-57-4
MDL Number: MFCD00003378
Molecular Formula: C7H5FO
Molecular Weight: 124.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 56
Specific Gravity (20/20): 1.18

Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 181 °C
Melting Pt: -10 °C
Density: 1.18 (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003378
CAS Number: 459-57-4
EINECS: 207-293-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3ad4879cca545b0fefe2450c720747d5452dea5&vid=50c64be518a1ece4be32ce0ba69aa98db7b7fb00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR