2-Fluorobenzaldehyde ≥98.0%

Supplier: TCI America

F0190-25ML F0190-250ML
TCF0190-025MLEA 25 USD
TCF0190-025ML TCF0190-250ML
2-Fluorobenzaldehyde ≥98.0%
2-Fluorobenzaldehyde

CAS Number: 446-52-6
MDL Number: MFCD00003302
Molecular Formula: C7H5FO
Molecular Weight: 124.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Melting point (°C): -45
Flash Point (°C): 55
Specific Gravity (20/20): 1.19

Formula: C₇H₅FO
Boiling Pt: 175 °C
Melting Pt: –45 °C
Density: 1.19 (20 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00003302
CAS Number: 446-52-6
EINECS: 207-171-2
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b80e2b9ea627d3626efaab409c1080d77d5f3101&vid=4a77ed761a2f34f86389e938a7d0235da0ccaeaf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR