4-Formylmorpholine ≥99.0%

Supplier: TCI America

Synonyms: Morpholine-4-carbaldehyde, N-Formylmorpholine

F0157-25G F0157-500G
TCF0157-025GEA 22.39 USD
TCF0157-025G TCF0157-500G
4-Formylmorpholine ≥99.0%
4-Formylmorpholine

CAS Number: 4394-85-8
MDL Number: MFCD00006170
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 237
Flash Point (°C): 125
Freezing point (°C): 23

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 237 °C
Flash Pt: 125 °C
MDL Number: MFCD00006170
CAS Number: 4394-85-8
EINECS: 224-518-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4547bee7492687e6042a205fba588494d3dca5de&vid=4547bee7492687e6042a205fba588494d3dca5de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR