1-Fluoro-4-nitrobenzene ≥98.0%

Supplier: TCI America

Synonyms: 4-Fluoronitrobenzene, 1-Fluoro-4-nitrobenzene

F0105-500G F0105-25G F0105-100G
TCF0105-500GEA 132.23 USD
TCF0105-500G TCF0105-25G TCF0105-100G
1-Fluoro-4-nitrobenzene ≥98.0%
1-Fluoro-4-nitrobenzene

CAS Number: 350-46-9
MDL Number: MFCD00007282
Molecular Formula: C6H4FNO2
Molecular Weight: 141.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 207
Flash Point (°C): 98
Freezing point (°C): 21

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 207 °C
Melting Pt: 21 °C
Density: 1.326 g/cm³ (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007282
CAS Number: 350-46-9
EINECS: 206-502-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b26ce6aefeffe965d08bd599fe8da09322096e30&vid=b7e1181995ec1c90c7082cbbc3f65eac3ad8f9f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR