1-Fluoro-2-nitrobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
o-Fluoronitrobenzene, 2-Fluoronitrobenzene
F0104-500G
F0104-25G
F0104-100G
TCF0104-500GEA
95
USD
TCF0104-500G
TCF0104-25G
TCF0104-100G
1-Fluoro-2-nitrobenzene ≥98.0%
1-Fluoro-2-nitrobenzene
CAS Number: 1493-27-2
MDL Number: MFCD00007048
Molecular Formula: C6H4FNO2
Molecular Weight: 141.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 94
Specific Gravity (20/20): 1.34
Formula:
C₆H₄FNO₂ MW: 141.1 g/mol Boiling Pt: 222 °C Melting Pt: –8 °C Density: 1.34 (20 °C) Flash Pt: 94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007048 CAS Number: 1493-27-2 EINECS: 216-088-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...