3-Fluoroaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
F0101-250G
F0101-25G
TCF0101-250GEA
243
USD
TCF0101-250G
TCF0101-25G
3-Fluoroaniline ≥98.0% (by GC, titration analysis)
3-Fluoroaniline
CAS Number: 372-19-0
MDL Number: MFCD00007758
Molecular Formula: C6H6FN
Molecular Weight: 111.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 186
Flash Point (°C): 77
Specific Gravity (20/20): 1.16
Formula:
C₆H₆FN MW: 111.12 g/mol Boiling Pt: 186 °C Melting Pt: –2 °C Density: 1.16 (20 °C) Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007758 CAS Number: 372-19-0 EINECS: 206-747-0 UN: 2941 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...