Furan ≥99.0% stabilized

Supplier: TCI America
Danger


Stabilization: Stabilised with Stabilised

F0074-25ML F0074-500ML
TCF0074-25MLEA 32.05 USD
TCF0074-25ML TCF0074-500ML
Furan ≥99.0% stabilized
Furan

(stabilized with BHT)
CAS Number: 110-00-9
MDL Number: MFCD00003222
Molecular Formula: C4H4O
Molecular Weight: 68.08
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 31
Melting point (°C): -84
Flash Point (°C): -35
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Divinylene Oxide, Oxol

Formula: C₄H₄O
Boiling Pt: 31 °C
Melting Pt: –84 °C
Density: 0.94 (20 °C)
Flash Pt: –35 °C
MDL Number: MFCD00003222
CAS Number: 110-00-9
EINECS: 203-727-3
UN: 2389
ADR: 3,I
Merck Index: 13,04317

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR