Fumaric acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 110-17-8
MDL Number: MFCD00002700
Molecular Formula: C4H4O4
Molecular Weight: 116.07
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 287
Other Product Names / Synonyms: trans-2-Butenedioic Acid
Formula:
C₄H₄O₄ Boiling Pt: 356 °C (1013 hPa) Melting Pt: 287 °C Density: 1.625 g/cm³ (20 °C) Flash Pt: 273 °C |
MDL Number:
MFCD00002700 CAS Number: 110-17-8 EINECS: 203-743-0 Merck Index: 13,04308 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...