Propyl formate ≥95.0%
Supplier: TCI America
|
Danger
|
F0058-500ML
F0058-25ML
TCF0058-500MLEA
108
USD
TCF0058-500ML
TCF0058-25ML
Propyl formate ≥95.0%
Propyl formate
CAS Number: 110-74-7
MDL Number: MFCD00003295
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 0
Specific Gravity (20/20): 0.90
Other Product Names / Synonyms: Formic Acid Propyl Ester
Formula:
C₄H₈O₂ Boiling Pt: 81 °C Melting Pt: -93 °C Density: 0.90 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003295 CAS Number: 110-74-7 EINECS: 203-798-0 UN: 1281 ADR: 3,II Merck Index: 12,08043 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...