Methyl formate ≥95.0%

Supplier: TCI America
Danger

F0057-25ML F0057-500ML
TCF0057-25MLEA 18 USD
TCF0057-25ML TCF0057-500ML
Methyl formate ≥95.0%
Methyl formate

CAS Number: 107-31-3
MDL Number: MFCD00003291
Molecular Formula: C2H4O2
Molecular Weight: 60.05
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 32
Flash Point (°C): -20
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Formic Acid Methyl Ester

Formula: C₂H₄O₂
Boiling Pt: 32 °C
Melting Pt: –100.4 °C
Density: 0.97 (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003291
CAS Number: 107-31-3
EINECS: 203-481-7
UN: 1243
ADR: 3,I
Merck Index: 12,06153
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=230bf9495466b17e7820737beef0a2a0657ff670&vid=230bf9495466b17e7820737beef0a2a0657ff670&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR