4-Fluorotoluene ≥99.0%

Supplier: TCI America

Synonyms: 1-Fluoro-4-methylbenzene

F0041-500ML F0041-25ML
TCF0041-500MLEA 276 USD
TCF0041-500ML TCF0041-25ML
4-Fluorotoluene ≥99.0%
4-Fluorotoluene

CAS Number: 352-32-9
MDL Number: MFCD00000358
Molecular Formula: C7H7F
Molecular Weight: 110.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Melting point (°C): -56
Flash Point (°C): 10
Specific Gravity (20/20): 1.00

Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 116 °C
Melting Pt: –56 °C
Density: 1.00 (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000358
CAS Number: 352-32-9
EINECS: 206-520-6
UN: 2388
ADR: 3,II
Merck Index: 13,04207
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f502a0d6dc7f92e7324a10e4d424f7c45d0f18b&vid=08b042d4bbe60044a35bd454c05ce2d8d0600a9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR