3-Fluorotoluene ≥99.0%

Supplier: TCI America

F0040-500ML F0040-25ML
TCF0040-500MLEA 99 USD
TCF0040-500ML TCF0040-25ML
3-Fluorotoluene ≥99.0%
3-Fluorotoluene

CAS Number: 352-70-5
MDL Number: MFCD00000339
Molecular Formula: C7H7F
Molecular Weight: 110.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 9
Specific Gravity (20/20): 1.00

Formula: C₇H₇F
Boiling Pt: 115 °C
Melting Pt: –88 °C
Density: 1.00 (20 °C)
Flash Pt: 9 °C
Storage Temperature: Ambient
MDL Number: MFCD00000339
CAS Number: 352-70-5
EINECS: 206-524-8
UN: 2388
ADR: 3,II
Merck Index: 13,04207
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6c2d83a89da4d88ba15339c0de3485e6d42170d&vid=98d8491353cc91829014ed2ce7df56b9e82f40e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR