2-Fluorobenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
F0035-500G
F0035-25G
TCF0035-500GEA
1119
USD
TCF0035-500G
TCF0035-025G
2-Fluorobenzoic acid ≥98.0% (by titrimetric analysis)
2-Fluorobenzoic acid
CAS Number: 445-29-4
MDL Number: MFCD00002405
Molecular Formula: C7H5FO2
Molecular Weight: 140.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 126
Formula:
C₇H₅FO₂ Boiling Pt: 245 °C (1013 hPa) Melting Pt: 126 °C Density: 1.46 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002405 CAS Number: 445-29-4 EINECS: 207-158-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...