Fluorobenzene ≥99.0%
Supplier: TCI America
F0034-500G
F0034-25G
F0034-100G
TCF0034-500GEA
121.44
USD
TCF0034-500G
TCF0034-25G
TCF0034-100G
Fluorobenzene ≥99.0%
Fluorobenzene
CAS Number: 462-06-6
MDL Number: MFCD00000280
Molecular Formula: C6H5F
Molecular Weight: 96.10
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): -13
Specific Gravity (20/20): 1.03
Formula:
C₆H₅F Boiling Pt: 85 °C Melting Pt: –41 °C Density: 1.03 (20 °C) Flash Pt: –13 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000280 CAS Number: 462-06-6 EINECS: 207-321-7 UN: 2387 ADR: 3,II Merck Index: 13,04197 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...