2-Fluoroaniline ≥99.0%
Supplier: TCI America
|
Warning
|
F0032-500G
F0032-25G
F0032-100G
TCF0032-500GEA
125
USD
TCF0032-500G
TCF0032-25G
TCF0032-100G
2-Fluoroaniline ≥99.0%
2-Fluoroaniline
CAS Number: 348-54-9
MDL Number: MFCD00007642
Molecular Formula: C6H6FN
Molecular Weight: 111.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 60
Specific Gravity (20/20): 1.16
Formula:
C₆H₆FN MW: 111.12 g/mol Boiling Pt: 170 °C Melting Pt: –29 °C Density: 1.16 (20 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007642 CAS Number: 348-54-9 EINECS: 206-478-9 UN: 2941 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...