9-Fluorenone ≥98.0%

Supplier: TCI America

Synonyms: 9H-Fluoren-9-one

F0021-25G F0021-500G F0021-100G
TCF0021-025GEA 23.33 USD
TCF0021-025G TCF0021-500G TCF0021-100G
9-Fluorenone ≥98.0%
9-Fluorenone

CAS Number: 486-25-9
MDL Number: MFCD00001141
Molecular Formula: C13H8O
Molecular Weight: 180.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 342
Melting point (°C): 84
Flash Point (°C): 175

Formula: C₁₃H₈O
MW: 180.21 g/mol
Boiling Pt: 342 °C
Melting Pt: 84 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00001141
CAS Number: 486-25-9
EINECS: 207-630-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83bfcb39f9d00c2a1ae6bb6949621b8115ab3cd9&vid=83bfcb39f9d00c2a1ae6bb6949621b8115ab3cd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR