1-Ethyl-4-piperidone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
E0498-250ML
E0498-25ML
E0498-100ML
TCE0498-250MLEA
589
USD
TCE0498-250ML
TCE0498-025ML
76220-008
1-Ethyl-4-piperidone ≥98.0% (by GC, titration analysis)
1-Ethyl-4-piperidone
CAS Number: 3612-18-8
MDL Number: MFCD00013356
Molecular Formula: C7H13NO
Molecular Weight: 127.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 68
Specific Gravity (20/20): 0.96
Formula:
C₇H₁₃NO Boiling Pt: 80 °C Density: 0.96 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00013356 CAS Number: 3612-18-8 EINECS: 222-781-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...