(±)-1-Ethoxy-2-propanol ≥90.0%

Supplier: TCI America
Danger

Synonyms: 1-Ethoxy-2-propanol, (±)-Monopropylene glycol ethyl ether

E0446-25ML E0446-500ML
TCE0446-025MLEA 26.45 USD
TCE0446-025ML TCE0446-500ML
(±)-1-Ethoxy-2-propanol ≥90.0%
(±)-1-Ethoxy-2-propanol

CAS Number: 1569-02-4
MDL Number: MFCD00067050
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 38
Freezing point (°C): -90
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Propylene Glycol Monoethyl Ether

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 132 °C
Melting Pt: –59 °C
Density: 0.90 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00067050
CAS Number: 1569-02-4
EINECS: 216-374-5
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e927f43496574efeabb6678ee6f4a31ffc074cc&vid=3e927f43496574efeabb6678ee6f4a31ffc074cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR