N-Ethylbenzylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: N-Benzylethylamine, N-Benzylethanamine

E0383-500ML E0383-25ML
TCE0383-500MLEA 355.7 USD
TCE0383-500ML TCE0383-25ML
N-Ethylbenzylamine ≥97.0% (by GC, titration analysis)
N-Ethylbenzylamine

CAS Number: 14321-27-8
MDL Number: MFCD00009031
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 69
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: N-Benzylethylamine

Formula: C₆H₅CH₂NHCH₂CH₃
MW: 135.21 g/mol
Boiling Pt: 199 °C
Density: 0.92 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00009031
CAS Number: 14321-27-8
EINECS: 238-265-1
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a11b587097134aae031a3e6eb12c1c1f6ab13a3&vid=7a11b587097134aae031a3e6eb12c1c1f6ab13a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR