2-Ethyl-2-oxazoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
E0373-500G
E0373-25G
E0373-100G
TCE0373-500GEA
325
USD
TCE0373-500G
TCE0373-25G
TCE0373-100G
2-Ethyl-2-oxazoline ≥98.0% (by GC, titration analysis)
2-Ethyl-2-oxazoline
CAS Number: 10431-98-8
MDL Number: MFCD00005302
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 128
Flash Point (°C): 32
Specific Gravity (20/20): 0.98
Formula:
C₅H₉NO MW: 99.13 g/mol Boiling Pt: 128 °C Density: 0.98 (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00005302 CAS Number: 10431-98-8 EINECS: 233-912-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...