1-Ethyl-2-pyrrolidone ≥98.0%

Supplier: TCI America
Danger

Synonyms: 1-Ethyl-2-pyrrolidinone, N-Ethyl-2-pyrrolidone, 1-Ethylpyrrolidin-2-one

E0358-250ML E0358-25ML
TCE0358-250MLEA 193 USD
TCE0358-250ML TCE0358-025ML
1-Ethyl-2-pyrrolidone ≥98.0%
1-Ethyl-2-pyrrolidone

CAS Number: 2687-91-4
MDL Number: MFCD00003199
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 218
Flash Point (°C): 76
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: N-Ethyl-2-azapentanone, NEP

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 218 °C
Density: 1.00 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00003199
CAS Number: 2687-91-4
EINECS: 220-250-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a11315fa3c152bb352f16217ef767af41c0ae7f&vid=9a11315fa3c152bb352f16217ef767af41c0ae7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR