1-Ethylpiperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0355-500ML E0355-25ML E0355-100ML
TCE0355-500MLEA 130 USD
TCE0355-500ML TCE0355-25ML TCE0355-100ML
1-Ethylpiperazine ≥98.0% (by GC, titration analysis)
1-Ethylpiperazine

CAS Number: 5308-25-8
MDL Number: MFCD00059912
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 43
Specific Gravity (20/20): 0.90

Formula: C₆H₁₄N₂
Boiling Pt: 158 °C
Melting Pt: –60 °C
Density: 0.90 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00059912
CAS Number: 5308-25-8
EINECS: 226-166-6
UN: 2733
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d074c4620eb0b26218b127fe18fc6d509c29908&vid=5d074c4620eb0b26218b127fe18fc6d509c29908&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR