N-Ethylethylenediamine ≥99.0%

Supplier: TCI America

Synonyms: 2-Aminoethyl(ethyl)amine

E0294-500ML E0294-25ML E0294-100ML
TCE0294-500MLEA 396 USD
TCE0294-500ML TCE0294-25ML TCE0294-100ML
N-Ethylethylenediamine ≥99.0%
N-Ethylethylenediamine

CAS Number: 110-72-5
MDL Number: MFCD00008166
Molecular Formula: C4H12N2
Molecular Weight: 88.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 33
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: 2-Ethylaminoethylamine

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 130 °C
Density: 0.84 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00008166
CAS Number: 110-72-5
EINECS: 203-795-4
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=818c28cfe9ca6cdbae2476f3628dbc16a7f604c2&vid=818c28cfe9ca6cdbae2476f3628dbc16a7f604c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR