Ethyl vinyl ether ≥98.0% stabilised
Supplier: TCI America
Stabilization:
Stabilised with Stabilised
E0193-500ML
E0193-25ML
E0193-100ML
TCE0193-500MLEA
54.71
USD
TCE0193-500ML
TCE0193-25ML
TCE0193-100ML
Ethyl vinyl ether ≥98.0% stabilised
Ethyl vinyl ether
(stabilized with KOH)
CAS Number: 109-92-2
MDL Number: MFCD00009248
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 36
Melting point (°C): -115
Flash Point (°C): -45
Specific Gravity (20/20): 0.75
Storage Temperature: 0-10°C
Formula:
C₄H₈O Boiling Pt: 36 °C Melting Pt: –115 °C Density: 0.75 (20 °C) Flash Pt: –45 °C |
MDL Number:
MFCD00009248 CAS Number: 109-92-2 EINECS: 203-718-4 UN: 1302 ADR: 3,I |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...