2-Ethylpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
E0168-500ML
E0168-25ML
E0168-100ML
TCE0168-500MLEA
623
USD
TCE0168-500ML
TCE0168-25ML
TCE0168-100ML
2-Ethylpyridine ≥98.0% (by GC, titration analysis)
2-Ethylpyridine
CAS Number: 100-71-0
MDL Number: MFCD00006361
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 144
Melting point (°C): -63
Flash Point (°C): 29
Specific Gravity (20/20): 0.93
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 144 °C Melting Pt: –63 °C Density: 0.93 (20 °C) Flash Pt: 29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006361 CAS Number: 100-71-0 EINECS: 202-881-9 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...