1-Ethylpiperidine ≥98.0%

Supplier: TCI America
Danger

E0166-25ML E0166-500ML
TCE0166-25MLEA 39.69 USD
TCE0166-25ML TCE0166-500ML
1-Ethylpiperidine ≥98.0%
1-Ethylpiperidine

CAS Number: 766-09-6
MDL Number: MFCD00006507
Molecular Formula: C7H15N
Molecular Weight: 113.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 19
Specific Gravity (20/20): 0.83

Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 131
Melting Pt: -57
Density: 0.83 (20 °C)
Flash Pt: 19 °C
Storage Temperature: Ambient
MDL Number: MFCD00006507
CAS Number: 766-09-6
EINECS: 212-161-6
UN: 2386
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef0cdb5983294079ad0cd8c79ce311dd352b7dd9&vid=ef0cdb5983294079ad0cd8c79ce311dd352b7dd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR