4-Ethylphenol ≥97.0%
Supplier: TCI America
|
Warning
|
E0159-25G
E0159-500G
TCE0159-025GEA
24
USD
TCE0159-025G
TCE0159-500G
4-Ethylphenol ≥97.0%
4-Ethylphenol
CAS Number: 123-07-9
MDL Number: MFCD00002393
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 219
Flash Point (°C): 104
Freezing point (°C): 44
Formula:
C₈H₁₀O Boiling Pt: 219 °C Melting Pt: 40 - 43 °C Density: 1.011 g/cm³ (20 °C) Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002393 CAS Number: 123-07-9 EINECS: 204-598-6 UN: 2430 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...