2-Ethylhexylamine ≥98.0%

Supplier: TCI America
Danger

Synonyms: 2-Ethyl-1-hexylamine

E0126-500ML E0126-25ML E0126-100ML
TCE0126-500MLEA 41 USD
TCE0126-500ML TCE0126-25ML TCE0126-100ML
2-Ethylhexylamine ≥98.0%
2-Ethylhexylamine

CAS Number: 104-75-6
MDL Number: MFCD00008148
Molecular Formula: C8H19N
Molecular Weight: 129.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 53
Specific Gravity (20/20): 0.79


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Amino-2-ethylhexane, Octylamine

Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 168 °C
Melting Pt: –76 °C
Density: 0.79 (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00008148
CAS Number: 104-75-6
EINECS: 203-233-8
UN: 2276
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83b3aa9908b0bc816db05566b0989e82be747f0b&vid=83b3aa9908b0bc816db05566b0989e82be747f0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR