2-Ethylcaproic acid ≥99.0%

Supplier: TCI America
Warning

Synonyms: 2-Ethylhexanoic acid

E0120-25ML E0120-500ML
TCE0120-25MLEA 24.4 USD
TCE0120-25ML TCE0120-500ML
2-Ethylcaproic acid ≥99.0%
2-Ethylcaproic acid

CAS Number: 149-57-5
MDL Number: MFCD00002675
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 228
Flash Point (°C): 120
Specific Gravity (20/20): 0.91


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Octanoic Acid

Formula: C₈H₁₆O₂
Boiling Pt: 228 °C
Melting Pt: –82.8 °C
Density: 0.91 (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002675
CAS Number: 149-57-5
EINECS: 205-743-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01e32d8af2c10f46a61bcd5f310cc6f10ff96d10&vid=01e32d8af2c10f46a61bcd5f310cc6f10ff96d10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR