Ethylene glycol monoisopropyl ether ≥99.0%

Supplier: TCI America

Synonyms: 2-Isopropoxyethanol

E0110-25ML E0110-500ML
TCE0110-025MLEA 17 USD
TCE0110-025ML TCE0110-500ML
Ethylene glycol monoisopropyl ether ≥99.0%
Ethylene glycol monoisopropyl ether

CAS Number: 109-59-1
MDL Number: MFCD00002866
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Flash Point (°C): 46
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: 2-Isopropoxyethanol, Isopropyl Cellosolve, Isopropyl Glycol

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 141 °C
Density: 0.91 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00002866
CAS Number: 109-59-1
EINECS: 203-685-6
UN: 1992
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f81620b64ed7ba98e01f06a2768f0c1e6a17d48&vid=9f81620b64ed7ba98e01f06a2768f0c1e6a17d48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR