2-Ethyl-1-butanol ≥98.0%

Supplier: TCI America
Warning

E0068-25ML E0068-100ML
TCE0068-25MLEA 25 USD
TCE0068-25ML TCE0068-100ML
2-Ethyl-1-butanol ≥98.0%
2-Ethyl-1-butanol

CAS Number: 97-95-0
MDL Number: MFCD00004744
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Melting point (°C): -15
Flash Point (°C): 57
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: 2-Ethylbutyl Alcohol

Formula: C₆H₁₄O
Boiling Pt: 146 °C
Melting Pt: –15 °C
Density: 0.83 (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00004744
CAS Number: 97-95-0
EINECS: 202-621-4
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d843edb14d98e140b66e9ff79b4fa631861ce244&vid=d843edb14d98e140b66e9ff79b4fa631861ce244&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR