Ethylbenzene ≥99.0%

Supplier: TCI America
Danger

Synonyms: Phenylethane

E0064-500ML E0064-25ML
TCE0064-500MLEA 40.55 USD
TCE0064-500ML TCE0064-25ML
Ethylbenzene ≥99.0%
Ethylbenzene

CAS Number: 100-41-4
MDL Number: MFCD00011647
Molecular Formula: C8H10
Molecular Weight: 106.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 25
Specific Gravity (20/20): 0.87

Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 136 °C
Melting Pt: -95 °C
Density: 0.87 (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3,II
Merck Index: 13,03801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2836d611086b5b60227a4ef9c3b1e03131835549&vid=22fc297aa5000d7e3278522aa4b1b4bd689f1df0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR