2-Ethylaminoethanol ≥98.0%

Supplier: TCI America

Synonyms: N-Ethyl-2-aminoethanol, N-Ethyl-2-ethanolamin, 2-(Ethylamino)ethan-1-ol

E0057-500ML E0057-25ML
TCE0057-500MLEA 45 USD
TCE0057-500ML TCE0057-25ML
2-Ethylaminoethanol ≥98.0%
2-Ethylaminoethanol

CAS Number: 110-73-6
MDL Number: MFCD00002841
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 169
Melting point (°C): -90
Flash Point (°C): 71
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: N-Ethylethanolamine

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 167
Melting Pt: -9
Density: 0.92 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002841
CAS Number: 110-73-6
EINECS: 203-797-5
UN: 2922
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d2dc9519fd780b6d978402c2f1a855fea2f4880&vid=6d2dc9519fd780b6d978402c2f1a855fea2f4880&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR