2-Ethoxyethyl acetate ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Ethylglycol acetate
E0049-500ML
E0049-25ML
TCE0049-500MLEA
24
USD
TCE0049-500ML
TCE0049-25ML
2-Ethoxyethyl acetate ≥98.0%
2-Ethoxyethyl acetate
CAS Number: 111-15-9
MDL Number: MFCD00009251
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Melting point (°C): -62
Flash Point (°C): 55
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: Acetic Acid 2-Ethoxyethyl Ester, Cellosolve Acetate, Ethylene Glycol Monoethyl Ether Acetate, Ethyl Glycol Acetate
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 156 °C Melting Pt: –62 °C Density: 0.97 (20 °C) Flash Pt: 55 °C |
MDL Number:
MFCD00009251 CAS Number: 111-15-9 EINECS: 203-839-2 UN: 1172 ADR: 3,III Merck Index: 13,03787 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...