2-Ethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0044-500G E0044-25G
TCE0044-500GEA 261 USD
TCE0044-500G TCE0044-025G
2-Ethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Ethoxybenzoic acid

CAS Number: 134-11-2
MDL Number: MFCD00002438
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Freezing point (°C): 19
Specific Gravity (20/20): 1.18

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 25 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002438
CAS Number: 134-11-2
EINECS: 205-130-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c9f338ee73cb1dde4ffcbfe278ae028d555b924&vid=47ca9c3ab91c6ae3862d7274ba2aa2189ab7c462&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR