(±)-Epichlorhydrin ≥99.0%

Supplier: TCI America

Synonyms: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane

E0012-500G E0012-25G
TCE0012-500GEA 40.56 USD
TCE0012-500G TCE0012-25G
(±)-Epichlorhydrin ≥99.0%
(±)-Epichlorhydrin

CAS Number: 106-89-8
MDL Number: MFCD00005132
Molecular Formula: C3H5ClO
Molecular Weight: 92.52
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Melting point (°C): -26
Flash Point (°C): 32
Specific Gravity (20/20): 1.18


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Chloromethyloxirane, 3-Chloropropylene Oxide, 1-Chloro-2,3-epoxypropane

Formula: C₃H₅ClO
MW: 92.52 g/mol
MDL Number: MFCD00005132
CAS Number: 106-89-8
Merck Index: 13,03642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85b8aef6117b99e28f07bb92a0c1f628d5b81c4a&vid=d4976897766e7df5413d57e65182fef6440cd13a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR