2,4-Difluorophenylhydrazinium chloride ≥97.0%

Supplier: TCI America

Synonyms: 2,4-Difluorophenylhydrazine hydrochloride, 1-(2,4-Difluorophenyl)hydrazine hydrochloride

D2698-5G D2698-25G
TCD2698-005GEA 68 USD
TCD2698-005G TCD2698-25G
2,4-Difluorophenylhydrazinium chloride ≥97.0%
2,4-Difluorophenylhydrazinium chloride

CAS Number: 51523-79-6
MDL Number: MFCD00013383
Molecular Formula: C6H6F2N2
Molecular Weight: 180.58
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,4-Difluorohydrazinobenzene Hydrochloride

Formula: C₆H₇ClF₂N₂
MDL Number: MFCD00013383
CAS Number: 51523-79-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc81e18276125ec55f0e593878d58fff4e4e36f&vid=9cc81e18276125ec55f0e593878d58fff4e4e36f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR