2,3-Dichlorophenylhydrazinium chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,3-Dichlorophenylhydrazine hydrochloride, (2,3-Dichlorophenyl)hydrazine hydrochloride, 1-(2,3-Dichlorophenyl)hydrazine hydrochloride

D2641-5G D2641-25G
TCD2641-005GEA 198 USD
TCD2641-005G TCD2641-025G
2,3-Dichlorophenylhydrazinium chloride ≥97.0% (by titrimetric analysis)
2,3-Dichlorophenylhydrazinium chloride

CAS Number: 21938-47-6
MDL Number: MFCD00052265
Molecular Formula: C6H6Cl2N2
Molecular Weight: 213.49
Purity/Analysis Method: >97.0% (T)
Form: Crystal

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00052265
CAS Number: 21938-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4871293be81b710f4c1ab81be74805ebf93270a&vid=f4871293be81b710f4c1ab81be74805ebf93270a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR