N,N'-Bis(tert-butyl)ethylenediamine ≥98.0%

Supplier: TCI America

Synonyms: N1,N2-Di-tert-butylethane-1,2-diamine, N,N'-Di-tert-butylethylenediamine

D2638-500ML D2638-25ML
TCD2638-500MLEA 543 USD
TCD2638-500ML TCD2638-25ML
N,N'-Bis(tert-butyl)ethylenediamine ≥98.0%
N,N'-Bis(tert-butyl)ethylenediamine

CAS Number: 4062-60-6
MDL Number: MFCD00014996
Molecular Formula: C10H24N2
Molecular Weight: 172.32
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 62
Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: 1,2-Bis(tert-butylamino)ethane

Formula: C₁₀H₂₄N₂
MW: 172.31 g/mol
Boiling Pt: 198 °C
Density: 0.80 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00014996
CAS Number: 4062-60-6
EINECS: 223-769-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2edad7331c494dc566c447687a764d8db5a2ef76&vid=2edad7331c494dc566c447687a764d8db5a2ef76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR