3,5-Difluorobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

D2626-5G D2626-25G
TCD2626-005GEA 62.8 USD
TCD2626-005G TCD2626-25G
3,5-Difluorobenzaldehyde ≥98.0%
3,5-Difluorobenzaldehyde

CAS Number: 32085-88-4
MDL Number: MFCD00010329
Molecular Formula: C7H4F2O
Molecular Weight: 142.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 56
Freezing point (°C): 25

Formula: C₇H₄F₂O
Boiling Pt: 61 - 63 °C (43 -rr)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00010329
CAS Number: 32085-88-4
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98b1ca28dca986d77bcd7dd823e76b754ac687de&vid=145e4f3395cab58a376dd4c7a579fddc027e5949&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR