2,3-Difluorobenzaldehyde ≥97.0%

Supplier: TCI America
Warning

D2614-5G D2614-25G
TCD2614-005GEA 83.7 USD
TCD2614-005G TCD2614-25G
2,3-Difluorobenzaldehyde ≥97.0%
2,3-Difluorobenzaldehyde

CAS Number: 2646-91-5
MDL Number: MFCD00010292
Molecular Formula: C7H4F2O
Molecular Weight: 142.10
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 65
Flash Point (°C): 58
Specific Gravity (20/20): 1.30

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 65 °C
Density: 1.30 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00010292
CAS Number: 2646-91-5
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a225687c43628eda98df680185b0ee95679fabf&vid=192c57144cc8061bebb77c1409564bfa385bb5fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR